accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSN

CAS RN 19989-66-3

IUPAC Name: (1,3-benzothiazol-6-yl)methanol
Synonyms: 1,3-benzothiazol-6-ylmethanol
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: OZGXSRLIKDPNMX-UHFFFAOYSA-N
SMILES: OCC1=CC=C2N=CSC2=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results