Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(Hydroxymethyl)benzothiazole, 97%, Thermo Scientific™

Catalog No. AC441330010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441330010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC441330010 Supplier Thermo Scientific Chemicals Supplier No. 441330010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19989-66-3
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
InChI Key OZGXSRLIKDPNMX-UHFFFAOYSA-N
Synonym benzo d thiazol-6-ylmethanol, 6-hydroxymethyl benzothiazole, 6-benzothiazolemethanol, benzothiazol-6-yl-methanol, benzothiazol-6-yl methanol, benzothiazole-6-methanol, benzothiazol-6-ylmethanol, 6-hydroxymethylbenzothiazole, benzo d thiazol-6-yl-methanol
PubChem CID 17860385
IUPAC Name 1,3-benzothiazol-6-ylmethanol
SMILES C1=CC2=C(C=C1CO)SC=N2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 103.0°C to 105.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 1 g
Formula Weight 165.22
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 6-(Hydroxymethyl)benzothiazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.