accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHONHOCH3NH2

CAS RN 1999-42-4

IUPAC Name: 2-(2-aminopropanamido)-4-methylpentanoic acid
Synonyms: 2-(2-aminopropanamido)-4-methylpentanoic acid
Molecular Weight (g/mol): 202.25
Molecular Formula: C9H18N2O3
InChi Key: RDIKFPRVLJLMER-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(=O)C(C)N)C(O)=O
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)