Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Alanyl-DL-leucine 95.0+%, TCI America™
SDP

Catalog No. A0185100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0185100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0185100MG Supplier TCI America Supplier No. A0185100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dipeptide reagent for laboratory research applications with >95.0% purity.

Chemical Identifiers

CAS 1999-42-4
Molecular Formula C9H18N2O3
Molecular Weight (g/mol) 202.254
MDL Number MFCD00025555
InChI Key RDIKFPRVLJLMER-UHFFFAOYSA-N
Synonym H-DL-Ala-DL-Leu-OH
PubChem CID 259583
IUPAC Name 2-(2-aminopropanoylamino)-4-methylpentanoic acid
SMILES CC(C)CC(C(=O)O)NC(=O)C(C)N

Specifications

Color White
Quantity 100 mg
Formula Weight 202.25
Percent Purity ≥95.0% (T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material DL-Alanyl-DL-leucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.