accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 200004-38-2

IUPAC Name: 1-[bromobis(4-fluorophenyl)methyl]-4-fluorobenzene
Synonyms: 1-[bromobis(4-fluorophenyl)methyl]-4-fluorobenzene
Molecular Weight (g/mol): 377.2
Molecular Formula: C19H12BrF3
InChi Key: DYMCGODRYMABQY-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(Br)(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results