Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4',4″-Trifluorotrityl Bromide 97.0+%, TCI America™
SDP

Catalog No. T14651G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T14651G Supplier TCI America Supplier No. T14651G

Chemical Identifiers

CAS 200004-38-2
Molecular Formula C19H12BrF3
Molecular Weight (g/mol) 377.204
MDL Number MFCD00191586
InChI Key DYMCGODRYMABQY-UHFFFAOYSA-N
Synonym Tris(4-fluorophenyl)bromomethane
PubChem CID 626724
IUPAC Name 1-[bromo-bis(4-fluorophenyl)methyl]-4-fluorobenzene
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)F)(C3=CC=C(C=C3)F)Br)F

Specifications

Melting Point 109°C
Quantity 1 g
UN Number 3261
Formula Weight 377.20
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′,4′′-Trifluorotrityl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.