accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 200068-36-6

IUPAC Name: 2,5-dibromo-1,1,3,4-tetraphenyl-1H-silole
Synonyms: 2,5-dibromo-1,1,3,4-tetraphenylsilole
Molecular Weight (g/mol): 544.36
Molecular Formula: C28H20Br2Si
InChi Key: MXGIPLZPCLLOTQ-UHFFFAOYSA-N
SMILES: BrC1=C(C(=C(Br)[Si]1(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results