accessibility menu, dialog, popup

UserName

Viewing profile as user:
KKHOONNOHOOOOO

CAS RN 2001-94-7

IUPAC Name: dipotassium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
Synonyms: dipotassium edta
Molecular Weight (g/mol): 368.43
Molecular Formula: C10H14K2N2O8
InChi Key: QLBHNVFOQLIYTH-UHFFFAOYSA-L
SMILES: [K+].[K+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results