Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA, Dipotassium Salt Dihydrate, MP Biomedicals

Catalog No. ICN203867 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN203867 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN203867 Supplier MP Biomedicals, Inc Supplier No. 0520386710
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2001-94-7
Molecular Formula C10H16K2N2O8
Molecular Weight (g/mol) 370.441
InChI Key MGDKBCNOUDORNI-UHFFFAOYSA-N
Synonym dipotassium dihydrogen ethylenediaminetetraacetate
PubChem CID 87063472
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;potassium
SMILES C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.[K].[K]

Specifications

Quantity 10 g
Formula Weight 404.46
Chemical Name or Material EDTA
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.