accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3C(S)NHO(S)H3COONHO(R)OO(S)HNO(S)CH3OO(R)HNO(R)OO(S)HNO(S)CH3OO(R)NHO(R)OOH3CCH3H3CCH3H3CCH3CH3CH3CH3CH3CH3H3CCH3H3CCH3H3C

CAS RN 2001-95-8

IUPAC Name: (3S,6S,9R,12R,15S,18S,21R,24R,27S,30S,36R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nonakis(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexaazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
Synonyms: valinomycin
Molecular Weight (g/mol): 1111.34
Molecular Formula: C54H90N6O18
InChi Key: FCFNRCROJUBPLU-UYBNATROSA-N
SMILES: CC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)C(NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C
Physical Form 
  • (7)
  • (2)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
Type 
  • (7)
Color 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (5)
  • (2)
  • (2)
  • (7)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (7)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)