accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20031-21-4

IUPAC Name: (3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol
Synonyms: (3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
Molecular Weight (g/mol): 190.2
Molecular Formula: C8H14O5
InChi Key: JAUQZVBVVJJRKM-XZBKPIIZSA-N
SMILES: CC1(C)O[C@H]2O[C@H](CO)[C@H](O)[C@H]2O1
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (6)
  • (1)
  • (5)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)