Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Cyanoborohydride Solution

Catalog No. 296813100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-681-3100ML 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 29-681-3100ML Supplier Sigma Aldrich Supplier No. 296813100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium cyanoborohydride is a versatile reducing agent stable in aqueous solution at pH 7.2. It is a weaker reducing agent as compared to NaBH4. It effectively reduces Schiff bases. NaCNBH4 participates in the reductive methylation for the efficient and selective conversion of the e-amino groups of lysyl residues in proteins to the mono- and dimethyl derivatives.Sodium cyanoborohydride may be used in the following studies:As reducing agent for the in vitro radiolabeling of proteins by reductive alkylation.Reduction of aldehydes and ketones.Selective deoxygenation of aldehydes and ketones.Preparation of colloidal gold particles of varying sizes.

Specifications

CAS 20031-21-4
Melting Point 69°C to 71°C
Boiling Point 112°C to 114°C (0.06 mmHg, lit.)
Molecular Formula C8H14O5
Linear Formula C8H14O5
MDL Number MFCD00063295
Quantity 25 g
Synonym Sodium borocyanohydride; Sodium cyanoborohydride; Sodium cyanotrihydroborate
Molecular Weight (g/mol) 190.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.