accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOHONH(S)CH3H3C

CAS RN 200616-39-3

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-[(2-phenylpropan-2-yl)oxy]pentanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-oxo-5-[(2-phenylpropan-2-yl)oxy]pentanoic acid
Molecular Weight (g/mol): 487.55
Molecular Formula: C29H29NO6
InChi Key: NPNRKQDJIWMHNG-VWLOTQADSA-N
SMILES: CC(C)(OC(=O)CC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O)C1=CC=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)