accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONOOH3COH3C(E)

CAS RN 20069-09-4

IUPAC Name: 1-[(2E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoyl]-1,2,5,6-tetrahydropyridin-2-one
Synonyms: piperlongumine piplartine
Molecular Weight (g/mol): 317.34
Molecular Formula: C17H19NO5
InChi Key: VABYUUZNAVQNPG-BQYQJAHWSA-N
SMILES: COC1=CC(\C=C\C(=O)N2CCC=CC2=O)=CC(OC)=C1OC
Form 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results