Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperlongumine 97.0+%, TCI America™
SDP

Catalog No. P236125MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P236125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P236125MG Supplier TCI America Supplier No. P236125MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 20069-09-4
Melting Point 125°C
Color White-Yellow
Molecular Formula C17H19NO5
Quantity 25 mg
MDL Number MFCD00075706
Synonym piperlongumine, piplartine, piplartin, unii-sgd66v4svj, sgd66v4svj, 5,6-dihydro-1-2e-1-oxo-3-3,4,5-trimethoxyphenyl-2-propen-1-yl-2 1h-pyridinone, e-1-3-3,4,5-trimethoxyphenyl acryloyl-5,6-dihydropyridin-2 1h-one, 1-e-3-3,4,5-trimethoxyphenyl prop-2-enoyl-2,3-dihydropyridin-6-one, e-5,6-dihydro-1-1-oxo-3-3,4,5-trimethoxyphenyl-2-propenyl-2 1h-pyridinone, 1-2e-3-3,4,5-trimethoxyphenyl prop-2-enoyl-1,2,5,6-tetrahydropyridin-2-one
InChI Key VABYUUZNAVQNPG-BQYQJAHWSA-N
SMILES COC1=CC(\C=C\C(=O)N2CCC=CC2=O)=CC(OC)=C1OC
IUPAC Name 1-[(2E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoyl]-1,2,5,6-tetrahydropyridin-2-one
Molecular Weight (g/mol) 317.34
PubChem CID 637858
ChEBI CHEBI:8241
Formula Weight 317.34
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Piperlongumine
Show More Show Less
RTECSNumber UU7785850
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.