accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2008-41-5

IUPAC Name: N,N-bis(2-methylpropyl)(ethylsulfanyl)formamide
Synonyms: sutan butilate (substance) butilate stauffer R-1910 caswell no. 434A diisocarb sutar' 85 E genate epa pesticide chemical code 041405 C11H23nos
Molecular Weight (g/mol): 217.37
Molecular Formula: C11H23NOS
InChi Key: BMTAFVWTTFSTOG-UHFFFAOYSA-N
SMILES: CCSC(=O)N(CC(C)C)CC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results