accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OFFFOCH3OFFFOMn(E/Z)(E/Z)

CAS RN 20080-72-2

IUPAC Name: manganese(2+) bis(1,1,1-trifluoro-4-oxopent-2-en-2-olate)
Synonyms: manganese(2+) bis(1,1,1-trifluoro-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 361.1
Molecular Formula: C10H8F6MnO4
InChi Key: RBWOFBBSLRFJMK-UHFFFAOYSA-L
SMILES: [Mn++].CC(=O)C=C([O-])C(F)(F)F.CC(=O)C=C([O-])C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results