Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(trifluoro-2,4-pentanedionato)manganese(II) 98.0+%, TCI America™
SDP

Catalog No. T06855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T06855G Supplier TCI America Supplier No. T06855G

Chemical Identifiers

CAS 20080-72-2
Molecular Formula C10H8F6MnO4
Molecular Weight (g/mol) 361.10
MDL Number MFCD00059872
InChI Key RBWOFBBSLRFJMK-UHFFFAOYSA-L
Synonym Manganese(II) Trifluoroacetylacetonate, Trifluoroacetylacetono Manganese(II) Salt
PubChem CID 53384599
IUPAC Name manganese(2+) bis(1,1,1-trifluoro-4-oxopent-2-en-2-olate)
SMILES [Mn++].CC(=O)C=C([O-])C(F)(F)F.CC(=O)C=C([O-])C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 361.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(trifluoro-2,4-pentanedionato)manganese(II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.