accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20082-71-7

IUPAC Name: chlorodimethyl(2,3,4,5,6-pentafluorophenyl)silane
Synonyms: chlorodimethyl(2,3,4,5,6-pentafluorophenyl)silane
Molecular Weight (g/mol): 260.66
Molecular Formula: C8H6ClF5Si
InChi Key: PQRFRTCWNCVQHI-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)C1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results