Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodimethylpentafluorophenylsilane 98%, Thermo Scientific™

Catalog No. AC210130050
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC210130050 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC210130050 Supplier Thermo Scientific Chemicals Supplier No. 210130050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20082-71-7
Molecular Formula C8H6ClF5Si
Molecular Weight (g/mol) 260.66
MDL Number MFCD00000500
InChI Key PQRFRTCWNCVQHI-UHFFFAOYSA-N
Synonym pentafluorophenyldimethylchlorosilane, flophemesyl chloride, chlorodimethylpentafluorophenylsilane, chlorodimethyl perfluorophenyl silane, silane, chlorodimethyl pentafluorophenyl, chlorodimethyl pentafluorophenyl silane, benzene, 1-chlorodimethylsilyl-2,3,4,5,6-pentafluoro, chlorodimethyl 2,3,4,5,6-pentafluorophenyl silane, chloro dimethyl pentafluorophenyl silane, dimethylpentafluorophenylchlorosilane
PubChem CID 88361
IUPAC Name chlorodimethyl(2,3,4,5,6-pentafluorophenyl)silane
SMILES C[Si](C)(Cl)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.4464 to 1.4484 (20°C, 589nm)
Linear Formula C6F5Si(CH3)2Cl
Quantity 5 mL
Formula Weight 260.66
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Chlorodimethylpentafluorophenylsilane, 98%
Hazard Category MOISTURE SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.