CAS RN 20109-09-5
IUPAC Name:
1-(2-phenylethynyl)cyclohexan-1-ol
Synonyms:
1-(2-phenylethynyl)cyclohexan-1-ol
Molecular Weight (g/mol):
200.28
Molecular Formula:
C14H16O
InChi Key:
LBEMYSKBSYEVAG-UHFFFAOYSA-N
SMILES:
OC1(CCCCC1)C#CC1=CC=CC=C1
1
–
1
of
1
results
1
1-(Phenylethynyl)cyclohexanol, 99%, Thermo Scientific™
CAS: 20109-09-5 Molecular Formula: C14H16O Molecular Weight (g/mol): 200.281 MDL Number: MFCD00060883 InChI Key: LBEMYSKBSYEVAG-UHFFFAOYSA-N Synonym: 1-phenylethynyl-1-cyclohexanol,1-phenylethynyl cyclohexanol,1-2-phenylethynyl cyclohexan-1-ol,1-phenylethynylcyclohexanol,acmc-1cey1,1-phenylethynyl-cyclohexanol,1-phenylethynyl cyclohexanol #,cyclohexanol,1-2-phenylethynyl,1-2-phenylethynyl-1-cyclohexanol,1-2-phenyl-ethynyl-cyclohexan-1-ol PubChem CID: 593776 IUPAC Name: 1-(2-phenylethynyl)cyclohexan-1-ol SMILES: C1CCC(CC1)(C#CC2=CC=CC=C2)O