Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Phenylethynyl)cyclohexanol, 99%, Thermo Scientific™

Catalog No. AAB2180806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2180806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2180806 Supplier Thermo Scientific Chemicals Supplier No. B2180806

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20109-09-5
Molecular Formula C14H16O
Molecular Weight (g/mol) 200.281
MDL Number MFCD00060883
InChI Key LBEMYSKBSYEVAG-UHFFFAOYSA-N
Synonym 1-phenylethynyl-1-cyclohexanol, 1-phenylethynyl cyclohexanol, 1-2-phenylethynyl cyclohexan-1-ol, 1-phenylethynylcyclohexanol, acmc-1cey1, 1-phenylethynyl-cyclohexanol, 1-phenylethynyl cyclohexanol #, cyclohexanol,1-2-phenylethynyl, 1-2-phenylethynyl-1-cyclohexanol, 1-2-phenyl-ethynyl-cyclohexan-1-ol
PubChem CID 593776
IUPAC Name 1-(2-phenylethynyl)cyclohexan-1-ol
SMILES C1CCC(CC1)(C#CC2=CC=CC=C2)O

Specifications

Melting Point 60°C to 62°C
Quantity 5 g
Formula Weight 200.28
Percent Purity 99%
Chemical Name or Material 1-(Phenylethynyl)cyclohexanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.