accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2013-26-5

IUPAC Name: sodium 2-oxobutanoate
Synonyms: sodium 2-oxobutanoate
Molecular Weight (g/mol): 124.07
Molecular Formula: C4H5NaO3
InChi Key: SUAMAHKUSIHRMR-UHFFFAOYSA-M
SMILES: [Na+].CCC(=O)C([O-])=O
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results