Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium 2-oxobutyrate

Catalog No. K08755G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K08755G 5 g
K087525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K08755G Supplier Sigma Aldrich Supplier No. K08755G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Substrate for the determination of lactate dehydrogenase isoenzymes.

Specifications

CAS 2013-26-5
Molecular Formula C4H5NaO3
Linear Formula CH3CH2COCOONa
Quantity 5 g
MDL Number MFCD00064194
Synonym 2-Oxobutanoic acid sodium salt; 2-Oxobutyric acid sodium salt; Sodium alpha-ketobutyrate
Molecular Weight (g/mol) 124.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.