accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20142-87-4

IUPAC Name: 2-(2-nitrophenoxy)acetyl chloride
Synonyms: 2-(2-nitrophenoxy)acetyl chloride
Molecular Weight (g/mol): 215.59
Molecular Formula: C8H6ClNO4
InChi Key: AEWBGACGIKCIJH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1OCC(Cl)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results