Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrophenoxyacetyl Chloride 95.0+%, TCI America™
SDP

Catalog No. N023610G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N023610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N023610G Supplier TCI America Supplier No. N023610G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20142-87-4
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.589
MDL Number MFCD00059822
InChI Key AEWBGACGIKCIJH-UHFFFAOYSA-N
PubChem CID 11321874
IUPAC Name 2-(2-nitrophenoxy)acetyl chloride
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])OCC(=O)Cl

Specifications

Melting Point 43°C
Color Yellow
Quantity 10 g
UN Number 3261
Formula Weight 215.59
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrophenoxyacetyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.