accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 201532-01-6

SOHOOONH(S)H3CCH3

CAS RN 201532-01-6

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methyl-3-[(triphenylmethyl)sulfanyl]butanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-methyl-3-[(triphenylmethyl)sulfanyl]butanoic acid
Molecular Weight (g/mol): 613.77
Molecular Formula: C39H35NO4S
InChi Key: XSGMGAINOILNJR-DHUJRADRSA-N
SMILES: CC(C)(SC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)