accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2016-05-9

IUPAC Name: [bis(cyclohexyloxy)methyl]dimethylazanium
Synonyms: [bis(cyclohexyloxy)methyl]dimethylazanium
Molecular Weight (g/mol): 256.41
Molecular Formula: C15H30NO2
InChi Key: GCCBITPXNLHRFH-UHFFFAOYSA-O
SMILES: C[NH+](C)C(OC1CCCCC1)OC1CCCCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results