Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide Dicyclohexyl Acetal 99%, Thermo Scientific™

Catalog No. AC162090500
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC162090500 50 g
AC162090100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC162090500 Supplier Thermo Scientific Chemicals Supplier No. 162090500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2016-05-9
Molecular Formula C15H30NO2
Molecular Weight (g/mol) 256.41
InChI Key GCCBITPXNLHRFH-UHFFFAOYSA-O
Synonym n,n-dimethylformamide dicyclohexyl acetal, methanamine, 1,1-bis cyclohexyloxy-n,n-dimethyl, dicyclohexyloxymethyl dimethylamine, bis cyclohexyloxy methyl dimethylamine, 1,1-dicyclohexyloxytrimethylamine, 1,1-bis cyclohexyloxy trimethylamine, n-bis cyclohexyloxy methyl-n,n-dimethylamine #
PubChem CID 74838
IUPAC Name 1,1-dicyclohexyloxy-N,N-dimethylmethanamine
SMILES CN(C)C(OC1CCCCC1)OC2CCCCC2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 0.95
Linear Formula (CH3)2NCH(OC6H11)2
Quantity 50 g
Formula Weight 255.4
Chemical Name or Material N, N-Dimethylformamide dicyclohexyl acetal, 99%
Hazard Category MOISTURE SENSITIVE
EINECSNumber 217-947-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.