CAS RN 201608-14-2
CAS RN 201608-14-2
IUPAC Name:
(4S)-4-{[(1S)-1-{[(1S)-2-carboxy-1-{[2-oxo-4-(trifluoromethyl)-2H-chromen-7-yl]carbamoyl}ethyl]carbamoyl}-2-methylpropyl]carbamoyl}-4-[(2S)-3-carboxy-2-acetamidopropanamido]butanoic acid
Synonyms:
(4S)-4-{[(1S)-1-{[(1S)-2-carboxy-1-{[2-oxo-4-(trifluoromethyl)chromen-7-yl]carbamoyl}ethyl]carbamoyl}-2-methylpropyl]carbamoyl}-4-[(2S)-3-carboxy-2-acetamidopropanamido]butanoic acid
Molecular Weight (g/mol):
729.62
Molecular Formula:
C30H34F3N5O13
InChi Key:
GZDRODOYEFEHGG-NUDCOPPTSA-N
SMILES:
CC(C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(C)=O)C(=O)N[C@@H](CC(O)=O)C(=O)NC1=CC=C2C(OC(=O)C=C2C(F)(F)F)=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals N-Acetyl-Asp-Glu-Val-Asp-7-amino-4-(trifluoromethyl)coumarin
CAS: 201608-14-2 Molecular Formula: C30H34F3N5O13,C30H34F3N5O13 Molecular Weight (g/mol): 729.62 InChI Key: GZDRODOYEFEHGG-NUDCOPPTSA-N Synonym: Ac-DEVD-AFC IUPAC Name: (4S)-4-{[(1S)-1-{[(1S)-2-carboxy-1-{[2-oxo-4-(trifluoromethyl)-2H-chromen-7-yl]carbamoyl}ethyl]carbamoyl}-2-methylpropyl]carbamoyl}-4-[(2S)-3-carboxy-2-acetamidopropanamido]butanoic acid SMILES: CC(C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(C)=O)C(=O)N[C@@H](CC(O)=O)C(=O)NC1=CC=C2C(OC(=O)C=C2C(F)(F)F)=C1