accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOCl

CAS RN 2019-34-3

IUPAC Name: 3-(4-chlorophenyl)propanoic acid
Synonyms: 3-(4-chlorophenyl)propanoic acid
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: BBSLOKZINKEUCR-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results