accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20194-18-7

IUPAC Name: sodium phenylmethanolate
Synonyms: sodium phenylmethanolate
Molecular Weight (g/mol): 130.12
Molecular Formula: C7H7NaO
InChi Key: LFQULJPVXNYWAG-UHFFFAOYSA-N
SMILES: [Na+].[O-]CC1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results