Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium benzyloxide, 1M solution in benzyl alcohol, AcroSeal™

Catalog No. AC433551000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433551000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC433551000 Supplier Thermo Scientific Chemicals Supplier No. 433551000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 20194-18-7 ∣ C7H7NaO ∣ 130.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 203°C to 205°C
Chemical Name or Material Sodium benzyloxide
Color Colorless to Yellow
CAS 20194-18-7, 100-51-6
Physical Form Viscous Liquid
Quantity 100 mL
Molecular Formula C7H7NaO
Linear Formula C6H5CH2ONa
Flash Point 90°C
Synonym sodium phenylmethanolate, sodium benzyloxide, benzyloxysodium, sodium benzylate, naobn, sodium phenylmethoxide, sodium benzyloxide solution
InChI Key LFQULJPVXNYWAG-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C[O-].[Na+]
IUPAC Name sodium;phenylmethanolate
Molecular Weight (g/mol) 130.12
PubChem CID 23676753
Formula Weight 130.12
Grade Ungraded
Concentration or Composition (by Analyte or Components) 0.95 to 1.10M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.077
Density 1.0770g/mL
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.