accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOCH3SFFF

CAS RN 202289-38-1

IUPAC Name: bis(2-methoxyethyl)(trifluoro-λ⁴-sulfanyl)amine
Synonyms: bis(2-methoxyethyl)(trifluoro-λ⁴-sulfanyl)amine
Molecular Weight (g/mol): 221.24
Molecular Formula: C6H14F3NO2S
InChi Key: APOYTRAZFJURPB-UHFFFAOYSA-N
SMILES: COCCN(CCOC)S(F)(F)F
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)