accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNSSSSNNCH3

CAS RN 20231-01-0

IUPAC Name: 4-methylpiperazine-1-carbothioylsulfanyl 4-methylpiperazine-1-carbodithioate
Synonyms: 4-methylpiperazine-1-carbothioylsulfanyl 4-methylpiperazine-1-carbodithioate
Molecular Weight (g/mol): 350.58
Molecular Formula: C12H22N4S4
InChi Key: OOBDBFPYJXJEHC-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C(=S)SSC(=S)N1CCN(C)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results