accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20235-19-2

IUPAC Name: (2S,3R)-2,3,4,5-tetrahydroxypentanal
Synonyms: (2S,3R)-2,3,4,5-tetrahydroxypentanal
Molecular Weight (g/mol): 150.13
Molecular Formula: C5H10O5
InChi Key: PYMYPHUHKUWMLA-DEOSMSJNSA-N
SMILES: OCC(O)[C@@H](O)[C@H](O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results