accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 202463-68-1

IUPAC Name: 1-(2,4-dichlorophenyl)-5-(4-iodophenyl)-4-methyl-N-(morpholin-4-yl)-1H-pyrazole-3-carboxamide
Synonyms: 1-(2,4-dichlorophenyl)-5-(4-iodophenyl)-4-methyl-N-(morpholin-4-yl)pyrazole-3-carboxamide
Molecular Weight (g/mol): 557.21
Molecular Formula: C21H19Cl2IN4O2
InChi Key: AJFFBPZYXRNAIC-UHFFFAOYSA-N
SMILES: CC1=C(N(N=C1C(=O)NN1CCOCC1)C1=C(Cl)C=C(Cl)C=C1)C1=CC=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results