accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20248-86-6

IUPAC Name: 4,4'-bis(bromomethyl)-1,1'-biphenyl
Synonyms: 4,4'-bis(bromomethyl)-1,1'-biphenyl
Molecular Weight (g/mol): 340.06
Molecular Formula: C14H12Br2
InChi Key: HMUGRILXVBKBID-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(C=C1)C1=CC=C(CBr)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)