Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis(bromomethyl)biphenyl 95.0+%, TCI America™
SDP

Catalog No. B19215G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19215G 5 g
B19211G 1 g
B192125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B19215G Supplier TCI America Supplier No. B19215G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20248-86-6
Molecular Formula C14H12Br2
Molecular Weight (g/mol) 340.06
MDL Number MFCD00094419
InChI Key HMUGRILXVBKBID-UHFFFAOYSA-N
PubChem CID 621238
IUPAC Name 4,4'-bis(bromomethyl)-1,1'-biphenyl
SMILES BrCC1=CC=C(C=C1)C1=CC=C(CBr)C=C1

Specifications

Melting Point 171°C
Quantity 5 g
UN Number 1759
Formula Weight 340.06
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(bromomethyl)biphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.