accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COON(Z)

CAS RN 2025-40-3

IUPAC Name: ethyl (2Z)-2-cyano-3-phenylprop-2-enoate
Synonyms: ethyl (2Z)-2-cyano-3-phenylprop-2-enoate
Molecular Weight (g/mol): 201.23
Molecular Formula: C12H11NO2
InChi Key: KCDAMWRCUXGACP-FLIBITNWSA-N
SMILES: CCOC(=O)C(=C/C1=CC=CC=C1)\C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results