Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl alpha-Cyanocinnamate 98.0+%, TCI America™
SDP

Catalog No. C044825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C044825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C044825G Supplier TCI America Supplier No. C044825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2025-40-3
Molecular Formula C12H11NO2
Molecular Weight (g/mol) 201.225
MDL Number MFCD00009137
InChI Key KCDAMWRCUXGACP-FLIBITNWSA-N
Synonym 2-Cyano-3-phenylacrylic Acid Ethyl Ester, alpha-Cyanocinnamic Acid Ethyl Ester, Ethyl 2-Cyano-3-phenylacrylate
PubChem CID 5354392
IUPAC Name ethyl (Z)-2-cyano-3-phenylprop-2-enoate
SMILES CCOC(=O)C(=CC1=CC=CC=C1)C#N

Specifications

Melting Point 50°C
Color White
Boiling Point 188°C
Quantity 25 g
Formula Weight 201.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl alpha-Cyanocinnamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.