CAS RN 20255-70-3
IUPAC Name:
4-iodo-N-(4-iodophenyl)aniline
Synonyms:
4-iodo-N-(4-iodophenyl)aniline
Molecular Weight (g/mol):
421.02
Molecular Formula:
C12H9I2N
InChi Key:
SJLIWXXAQHNDHM-UHFFFAOYSA-N
SMILES:
IC1=CC=C(NC2=CC=C(I)C=C2)C=C1
1
–
1
of
1
results
1
Bis(4-iodophenyl)amine 98.0+%, TCI America™
CAS: 20255-70-3 Molecular Formula: C12H9I2N Molecular Weight (g/mol): 421.02 InChI Key: SJLIWXXAQHNDHM-UHFFFAOYSA-N PubChem CID: 12950430 IUPAC Name: 4-iodo-N-(4-iodophenyl)aniline SMILES: C1=CC(=CC=C1NC2=CC=C(C=C2)I)I