Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-iodophenyl)amine 98.0+%, TCI America™
SDP

Catalog No. B26855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26855G 5 g
B26851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26855G Supplier TCI America Supplier No. B26855G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20255-70-3
Molecular Formula C12H9I2N
Molecular Weight (g/mol) 421.02
InChI Key SJLIWXXAQHNDHM-UHFFFAOYSA-N
PubChem CID 12950430
IUPAC Name 4-iodo-N-(4-iodophenyl)aniline
SMILES C1=CC(=CC=C1NC2=CC=C(C=C2)I)I

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 5 g
Synonym 4,4′-Diiododiphenylamine
Formula Weight 421.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-iodophenyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.