accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20265-38-7

IUPAC Name: 2-methoxypyridin-3-amine
Synonyms: 2-methoxypyridin-3-amine
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: YXFAOWYMDGUFIQ-UHFFFAOYSA-N
SMILES: COC1=C(N)C=CC=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)