Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2-methoxypyridine 98.0+%, TCI America™
SDP

Catalog No. A24035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24035G 5 g
A240325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24035G Supplier TCI America Supplier No. A24035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20265-38-7
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD00833386
InChI Key YXFAOWYMDGUFIQ-UHFFFAOYSA-N
Synonym 3-amino-2-methoxypyridine, 2-methoxy-3-aminopyridine, 2-methoxy-pyridin-3-ylamine, 3-pyridinamine, 2-methoxy, 2-methoxy-3-pyridylamine, 2-methoxy-3-pyridinamine, 2-methoxypyridine-3-amine, pubchem1269, 2-methoxy pyridin-3-amine
PubChem CID 2737488
IUPAC Name 2-methoxypyridin-3-amine
SMILES COC1=C(C=CC=N1)N

Specifications

Melting Point 67°C
Boiling Point 118°C
Quantity 5 g
Formula Weight 124.14
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2-methoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.