accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH2N

CAS RN 20265-39-8

IUPAC Name: 2-methoxypyridin-4-amine
Synonyms: 2-methoxypyridin-4-amine
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: SVEQHRSELNPKJJ-UHFFFAOYSA-N
SMILES: COC1=CC(N)=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results