Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-methoxypyridine 98.0+%, TCI America™
SDP

Catalog No. A25435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25435G 5 g
A25431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25435G Supplier TCI America Supplier No. A25435G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20265-39-8
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD06738657
InChI Key SVEQHRSELNPKJJ-UHFFFAOYSA-N
PubChem CID 280891
IUPAC Name 2-methoxypyridin-4-amine
SMILES COC1=NC=CC(=C1)N

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 5 g
Formula Weight 124.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-methoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.