accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 202865-60-9

IUPAC Name: 1-bromo-2,5-difluoro-4-methoxybenzene
Synonyms: 1-bromo-2,5-difluoro-4-methoxybenzene
Molecular Weight (g/mol): 223.02
Molecular Formula: C7H5BrF2O
InChi Key: OOTXNQBDWFJMNN-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(Br)C=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results