Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,5-difluoroanisole 98.0+%, TCI America™
SDP

Catalog No. B30975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B30975G Supplier TCI America Supplier No. B30975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 202865-60-9
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.017
MDL Number MFCD00143258
InChI Key OOTXNQBDWFJMNN-UHFFFAOYSA-N
Synonym 4-bromo-2,5-difluoroanisole, 1-bromo-2,5-difluoro-4-methoxy-benzene, timtec-bb sbb037912, pubchem2653, acmc-1cm1k, benzene,1-bromo-2,5-difluoro-4-methoxy, benzene, 1-bromo-2,5-difluoro-4-methoxy
PubChem CID 2724983
IUPAC Name 1-bromo-2,5-difluoro-4-methoxybenzene
SMILES COC1=CC(=C(C=C1F)Br)F

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 5 g
Formula Weight 223.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,5-difluoroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.