accessibility menu, dialog, popup

UserName

Viewing profile as user:
FIBr

CAS RN 202865-72-3

IUPAC Name: 1-bromo-4-fluoro-2-iodobenzene
Synonyms: 1-bromo-4-fluoro-2-iodobenzene
Molecular Weight (g/mol): 300.9
Molecular Formula: C6H3BrFI
InChi Key: QDCBSSUCEUMLGC-UHFFFAOYSA-N
SMILES: FC1=CC(I)=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results